siRNA Details
| Gene Symbol: SIX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005413.3 | NP_005404.1 | 2 | 1608 | ||
| GenBank mRNA | AJ012611.1 | - | 2 | 1608 | ||
| EL952320.1 | - | 1 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIX3| Gene Symbol: SIX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005413.3 | NP_005404.1 | 2 | 1608 | ||
| GenBank mRNA | AJ012611.1 | - | 2 | 1608 | ||
| EL952320.1 | - | 1 | 333 | |||