siRNA Details
| Gene Symbol: Cyp2c12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031572.1 | NP_113760.1 | 1 | 152 | ||
| GenBank mRNA | BC089790.1 | - | 1 | 160 | 7371667 | |
| FQ219491.1 | - | 1 | 177 | |||
| J03786.1 | - | 1 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c12| Gene Symbol: Cyp2c12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031572.1 | NP_113760.1 | 1 | 152 | ||
| GenBank mRNA | BC089790.1 | - | 1 | 160 | 7371667 | |
| FQ219491.1 | - | 1 | 177 | |||
| J03786.1 | - | 1 | 152 | |||