siRNA Details
| Gene Symbol: Acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017075.2 | NP_058771.2 | 7, 8 | 803 | ||
| GenBank mRNA | D00512.1 | - | 7, 8 | 735 | ||
| D13921.1 | - | 7, 8 | 735 | |||
| FM071064.1 | - | 5, 6 | 591 | |||
| FM098848.1 | - | 2, 3 | 185 | |||

Acat1
Genomic Map