siRNA Details
| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172305.1 | NP_001165776.1 | 7 | 807 | ||
| NM_012713.3 | NP_036845.3 | 7 | 807 | |||
| GenBank mRNA | K03485.1 | - | 4 | 441 | ||
| M13706.1 | - | 5 | 817 | |||
| M19007.1 | - | 5 | 767 | |||
| X04440.1 | - | 5 | 1361 | |||

Prkcb
Genomic Map