siRNA Details
| Gene Symbol: Adm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012715.1 | NP_036847.1 | 4 | 583 | ||
| XM_008759687.1 | XP_008757909.1 | 5 | 2090 | |||
| GenBank mRNA | BC061775.1 | - | 4 | 579 | 5621783 | |
| D15069.1 | - | 4 | 565 | |||
| U15419.1 | - | 4 | 583 | |||

Adm
Genomic Map