siRNA Details
| Gene Symbol: Ampd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138876.1 | NP_620231.1 | 16 | 2194 | ||
| XM_006233060.2 | XP_006233122.1 | 15 | 2271 | |||
| XM_008761369.1 | XP_008759591.1 | 15 | 2194 | |||
| GenBank mRNA | FQ214856.1 | - | 9 | 1293 | ||
| FQ216456.1 | - | 6 | 745 | |||
| FQ224901.1 | - | 6 | 682 | |||
| J02811.1 | - | 16 | 2194 | |||

Ampd1
Genomic Map