siRNA Details
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012717.1 | NP_036849.1 | 14 | 1392 | ||
XM_006234438.2 | XP_006234500.1 | 14 | 1647 | |||
XM_006234439.2 | XP_006234501.1 | 13 | 1485 | |||
GenBank mRNA | L27487.1 | - | 14 | 1665 | ||
X70658.1 | - | 14 | 1392 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
