siRNA Details
| Gene Symbol: Andpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012718.2 | NP_036850.1 | 3 | 403 | ||
| XM_006235184.2 | XP_006235246.1 | 3 | 642 | |||
| GenBank mRNA | BC065311.1 | - | 1 | 3823 | ||
| M27901.1 | - | 3 | 387 | |||
| M58167.1 | - | 3 | 404 | |||

Andpro
Genomic Map