siRNA Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012720.1 | NP_036852.1 | 3 | 261 | ||
| XM_006244065.2 | XP_006244127.1 | 3 | 345 | |||
| XM_006244067.2 | XP_006244129.1 | 4 | 383 | |||
| XM_006244069.2 | XP_006244131.1 | 4 | 383 | |||
| XM_008766669.1 | XP_008764891.1 | 2 | 282 | |||
| GenBank mRNA | AF157498.1 | - | 3 | 254 | ||
| BC087694.1 | - | 2 | 215 | |||
| BC161856.1 | - | 4 | 297 | 7320248 | ||
| D28110.1 | - | 2 | 212 | |||
| D28111.1 | - | 2 | 212 | |||
| X87900.1 | - | 3 | 254 | |||
| X89637.1 | - | 3 | 261 | |||
| X89638.1 | - | 2 | 243 | |||
| X90402.1 | - | 3 | 267 | |||

Mobp
Genomic Map