siRNA Details
| Gene Symbol: Gip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019630.3 | NP_062604.1 | 2 | 104 | ||
| XM_006247177.2 | XP_006247239.1 | 1 | 326 | |||
| XM_008767947.1 | XP_008766169.1 | 2 | 146 | |||
| GenBank mRNA | D11439.1 | - | 1 | 76 | ||
| L08831.1 | - | 2 | 104 | |||
| M92916.1 | - | 1 | 2013 | |||
| X66724.1 | - | 2 | 165 | |||
| Z19564.1 | - | 2 | 104 | |||

Gip
Genomic Map