siRNA Details
| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012721.2 | NP_036853.2 | 1 | 185 | ||
| XM_008768851.1 | XP_008767073.1 | 1 | 375 | |||
| GenBank mRNA | X92070.1 | - | 1 | 66 | ||
| X97376.1 | - | 1 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2rx6| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012721.2 | NP_036853.2 | 1 | 185 | ||
| XM_008768851.1 | XP_008767073.1 | 1 | 375 | |||
| GenBank mRNA | X92070.1 | - | 1 | 66 | ||
| X97376.1 | - | 1 | 196 | |||