siRNA Details
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012722.1 | NP_036854.1 | 16 | 933 | ||
GenBank mRNA | BC085910.1 | - | 16 | 933 | 7097661 | |
M60647.1 | - | 16 | 910 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012722.1 | NP_036854.1 | 16 | 933 | ||
GenBank mRNA | BC085910.1 | - | 16 | 933 | 7097661 | |
M60647.1 | - | 16 | 910 |