siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012727.3 | NP_036859.2 | 5, 6 | 520 | ||
| XM_006254523.2 | XP_006254585.1 | 4, 5 | 462 | |||
| GenBank mRNA | BC128706.1 | - | 5, 6 | 520 | 7455982 | |
| M63334.1 | - | 414 | ||||
| M64757.1 | - | 3, 4 | 510 | |||
| S65840.1 | - | 4, 5 | 445 | |||

Camk4
Genomic Map