siRNA Details
| Gene Symbol: Prss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012729.1 | NP_036861.1 | 2 | 114 | ||
| GenBank mRNA | V01274.1 | - | 2 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss2| Gene Symbol: Prss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012729.1 | NP_036861.1 | 2 | 114 | ||
| GenBank mRNA | V01274.1 | - | 2 | 91 | ||