siRNA Details
| Gene Symbol: Prss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012729.1 | NP_036861.1 | 2 | 73 | ||
| GenBank mRNA | V01274.1 | - | 2 | 50 | ||
| Gene Symbol: LOC102554637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236386.2 | XP_006236448.1 | 2 | 73 | ||
| Gene Symbol: LOC103690254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762925.1 | XP_008761147.1 | 2 | 73 | ||
| Gene Symbol: LOC683849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749732.3 | XP_003749780.1 | 2 | 73 | ||
| Gene Symbol: Prss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012635.1 | NP_036767.1 | 2 | 85 | ||
| GenBank mRNA | FQ211333.1 | - | 2 | 88 | ||
| FQ226369.1 | - | 2 | 86 | |||
| FQ227841.1 | - | 2 | 86 | |||
| FQ233648.1 | - | 2 | 86 | |||
| FQ235029.1 | - | 2 | 86 | |||
| V01273.1 | - | 2 | 85 | |||

Prss2
Genomic Map