siRNA Details
| Gene Symbol: Hk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012734.1 | NP_036866.1 | 12 | 2289 | ||
| GenBank mRNA | J04526.1 | - | 12 | 2289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hk1| Gene Symbol: Hk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012734.1 | NP_036866.1 | 12 | 2289 | ||
| GenBank mRNA | J04526.1 | - | 12 | 2289 | ||