siRNA Details
| Gene Symbol: Slc12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270617.1 | NP_001257546.1 | 8, 9 | 1298 | ||
| NM_001270618.1 | NP_001257547.1 | 8, 9 | 1298 | |||
| XM_008762140.1 | XP_008760362.1 | 10, 11 | 1562 | |||
| XM_008762141.1 | XP_008760363.1 | 10, 11 | 1561 | |||
| XM_008762142.1 | XP_008760364.1 | 9, 10 | 1466 | |||
| XM_008762143.1 | XP_008760365.1 | 9, 10 | 1466 | |||
| XM_008762144.1 | XP_008760366.1 | 8, 9 | 1370 | |||
| GenBank mRNA | BC087017.1 | - | 8, 9 | 1340 | 7109809 | |
| EF577032.1 | - | 7, 8 | 1282 | |||
| JN579708.1 | - | 8, 9 | 1298 | |||
| U10096.1 | - | 8, 9 | 1298 | |||

Slc12a1
Genomic Map