siRNA Details
| Gene Symbol: PVR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017076.2 | NP_058772.2 | 3 | 659 | ||
| GenBank mRNA | BC085741.1 | - | 3 | 659 | 7191422 | |
| L12025.1 | - | 3 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PVR| Gene Symbol: PVR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017076.2 | NP_058772.2 | 3 | 659 | ||
| GenBank mRNA | BC085741.1 | - | 3 | 659 | 7191422 | |
| L12025.1 | - | 3 | 648 | |||