siRNA Details
| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019135.1 | NP_062008.1 | 6, 7 | 747 | ||
| XM_008764206.1 | XP_008762428.1 | 6, 7 | 1311 | |||
| GenBank mRNA | D42117.1 | - | 6, 7 | 747 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf8| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019135.1 | NP_062008.1 | 6, 7 | 747 | ||
| XM_008764206.1 | XP_008762428.1 | 6, 7 | 1311 | |||
| GenBank mRNA | D42117.1 | - | 6, 7 | 747 | ||