siRNA Details
| Gene Symbol: Apoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012737.1 | NP_036869.1 | 3 | 731 | ||
| GenBank mRNA | BC091159.1 | - | 3 | 706 | 7373033 | |
| FQ218620.1 | - | 3 | 732 | |||
| M00002.1 | - | 3 | 731 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apoa4| Gene Symbol: Apoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012737.1 | NP_036869.1 | 3 | 731 | ||
| GenBank mRNA | BC091159.1 | - | 3 | 706 | 7373033 | |
| FQ218620.1 | - | 3 | 732 | |||
| M00002.1 | - | 3 | 731 | |||