siRNA Details
| Gene Symbol: Adra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012739.3 | NP_036871.3 | 1 | 357 | ||
| GenBank mRNA | U79031.1 | - | 1 | 312 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adra2a| Gene Symbol: Adra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012739.3 | NP_036871.3 | 1 | 357 | ||
| GenBank mRNA | U79031.1 | - | 1 | 312 | ||
Set Membership: |
|