siRNA Details
| Gene Symbol: Cyp2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031543.1 | NP_113731.1 | 3 | 393 | ||
| XM_006230520.1 | XP_006230582.1 | 4 | 576 | |||
| GenBank mRNA | AF061442.1 | - | 3 | 381 | ||
| BC081774.1 | - | 3 | 395 | 7110096 | ||
| FQ209397.1 | - | 3 | 411 | |||
| FQ210815.1 | - | 3 | 414 | |||
| FQ211086.1 | - | 3 | 414 | |||
| FQ218528.1 | - | 1 | 32 | |||
| FQ218532.1 | - | 3 | 409 | |||
| FQ219736.1 | - | 3 | 411 | |||
| J02627.1 | - | 3 | 393 | |||

Cyp2e1
Genomic Map