siRNA Details
| Gene Symbol: Pld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033299.2 | NP_150641.2 | 18 | 2293 | ||
| XM_008767778.1 | XP_008766000.1 | 20 | 2299 | |||
| GenBank mRNA | AB003172.1 | - | 19 | 2257 | ||
| D88672.1 | - | 18 | 2293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pld2| Gene Symbol: Pld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033299.2 | NP_150641.2 | 18 | 2293 | ||
| XM_008767778.1 | XP_008766000.1 | 20 | 2299 | |||
| GenBank mRNA | AB003172.1 | - | 19 | 2257 | ||
| D88672.1 | - | 18 | 2293 | |||