siRNA Details
| Gene Symbol: Foxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012743.1 | NP_036875.1 | 3 | 819 | ||
| XM_006235139.2 | XP_006235201.1 | 2 | 694 | |||
| XM_006235140.2 | XP_006235202.1 | 2 | 946 | |||
| GenBank mRNA | L09647.1 | - | 3 | 819 | ||

Foxa2
Genomic Map