siRNA Details
| Gene Symbol: Foxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017077.2 | NP_058773.1 | 2 | 680 | ||
| GenBank mRNA | L09648.1 | - | 2 | 681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxa3| Gene Symbol: Foxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017077.2 | NP_058773.1 | 2 | 680 | ||
| GenBank mRNA | L09648.1 | - | 2 | 681 | ||