siRNA Details
Gene Symbol: Nppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031545.1 | NP_113733.1 | 1 | 174 | ||
GenBank mRNA | FQ228997.1 | - | 230 | |||
M25297.1 | - | 1 | 174 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031545.1 | NP_113733.1 | 1 | 174 | ||
GenBank mRNA | FQ228997.1 | - | 230 | |||
M25297.1 | - | 1 | 174 |