siRNA Details
Gene Symbol: Chrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031547.1 | NP_113735.1 | 2 | 553 | ||
XM_008762022.1 | XP_008760244.1 | 2 | 455 | |||
GenBank mRNA | M16409.1 | - | 1 | 374 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
