siRNA Details
| Gene Symbol: Chrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031547.1 | NP_113735.1 | 2 | 811 | ||
| XM_008762022.1 | XP_008760244.1 | 2 | 713 | |||
| GenBank mRNA | M16409.1 | - | 1 | 632 | ||
More Information
Set Membership: |
|

Chrm4
Genomic Map