siRNA Details
| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030994.2 | NP_112256.1 | 15 | 2323 | ||
| XM_008760735.1 | XP_008758957.1 | 15 | 1939 | |||
| GenBank mRNA | X52140.1 | - | 15 | 2347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga1| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030994.2 | NP_112256.1 | 15 | 2323 | ||
| XM_008760735.1 | XP_008758957.1 | 15 | 1939 | |||
| GenBank mRNA | X52140.1 | - | 15 | 2347 | ||