siRNA Details
| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030994.2 | NP_112256.1 | 27, 28 | 3787 | ||
| XM_008760735.1 | XP_008758957.1 | 26, 27 | 3193 | |||
| GenBank mRNA | FQ227807.1 | - | 8, 9 | 752 | ||
| X52140.1 | - | 27, 28 | 3811 | |||

Itga1
Genomic Map