siRNA Details
| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100635.1 | NP_001094105.1 | 5 | 919 | ||
| XM_006250030.2 | XP_006250092.1 | 6 | 1155 | |||
| GenBank mRNA | Y13972.1 | - | 4 | 844 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mr1| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100635.1 | NP_001094105.1 | 5 | 919 | ||
| XM_006250030.2 | XP_006250092.1 | 6 | 1155 | |||
| GenBank mRNA | Y13972.1 | - | 4 | 844 | ||