siRNA Details
| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100635.1 | NP_001094105.1 | 3 | 507 | ||
| XM_006250030.2 | XP_006250092.1 | 3 | 621 | |||
| XM_008769606.1 | XP_008767828.1 | 3 | 621 | |||
| GenBank mRNA | Y13972.1 | - | 2 | 432 | ||

Mr1
Genomic Map