siRNA Details
| Gene Symbol: Myo5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017083.1 | NP_058779.1 | 6 | 814 | ||
| XM_006254908.2 | XP_006254970.1 | 2 | 267 | |||
| XM_008772085.1 | XP_008770307.1 | 6 | 777 | |||
| XM_008772086.1 | XP_008770308.1 | 6 | 777 | |||
| GenBank mRNA | U60416.1 | - | 6 | 814 | ||

Myo5b
Genomic Map