siRNA Details
Gene Symbol: Pcdhb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173099.1 | NP_775122.1 | 1 | 1075 | ||
XM_006254626.2 | XP_006254688.1 | 2 | 1660 | |||
GenBank mRNA | L43592.1 | - | 1 | 1075 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcdhb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173099.1 | NP_775122.1 | 1 | 1075 | ||
XM_006254626.2 | XP_006254688.1 | 2 | 1660 | |||
GenBank mRNA | L43592.1 | - | 1 | 1075 |