siRNA Details
| Gene Symbol: Pcdhb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173099.1 | NP_775122.1 | 1 | 206 | ||
| XM_006254626.2 | XP_006254688.1 | 2 | 791 | |||
| XM_006254627.2 | XP_006254689.1 | 1 | 698 | |||
| GenBank mRNA | L43592.1 | - | 1 | 206 | ||

Pcdhb12
Genomic Map