siRNA Details
| Gene Symbol: Ncl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012749.2 | NP_036881.2 | 12 | 1936 | ||
| GenBank mRNA | BC085751.1 | - | 12 | 1936 | 7109097 | |
| FJ817497.1 | - | 12 | 1375 | |||
| FQ219308.1 | - | 5 | 548 | |||
| FQ222026.1 | - | 2 | 182 | |||

Ncl
Genomic Map