siRNA Details
| Gene Symbol: Gfra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012750.1 | NP_036882.1 | 5, 6 | 1028 | ||
| XM_006252238.2 | XP_006252300.1 | 4, 5 | 1797 | |||
| GenBank mRNA | AF003825.1 | - | 1578 | |||
| AF005226.1 | - | 5, 6 | 908 | |||
| U97143.1 | - | 5, 6 | 1028 | |||

Gfra2
Genomic Map