siRNA Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012751.1 | NP_036883.1 | 7, 8 | 1061 | ||
| XM_006246596.2 | XP_006246658.1 | 8, 9 | 1034 | |||
| GenBank mRNA | BC085757.1 | - | 7, 8 | 1106 | 7192278 | |
| D28561.1 | - | 7, 8 | 1061 | |||
| J04524.1 | - | 7, 8 | 1043 | |||
| M25482.1 | - | 7, 8 | 1026 | |||
| X14771.1 | - | 7, 8 | 1074 | |||

Slc2a4
Genomic Map