siRNA Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012751.1 | NP_036883.1 | 11 | 1627 | ||
| XM_006246596.2 | XP_006246658.1 | 12 | 1600 | |||
| GenBank mRNA | BC085757.1 | - | 11 | 1672 | 7192278 | |
| D28561.1 | - | 11 | 1627 | |||
| J04524.1 | - | 11 | 1609 | |||
| M25482.1 | - | 11 | 1592 | |||
| X14771.1 | - | 11 | 1640 | |||

Slc2a4
Genomic Map