siRNA Details
| Gene Symbol: Esr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012754.1 | NP_036886.1 | 2, 3 | 636 | ||
| XM_006240221.2 | XP_006240283.1 | 2, 3 | 877 | |||
| GenBank mRNA | AB012721.1 | - | 1, 2 | 220 | ||
| AB190769.1 | - | 1, 2 | 459 | |||
| AB190770.1 | - | 1, 2 | 459 | |||
| AF042058.1 | - | 1, 2 | 220 | |||
| AF042059.1 | - | 1, 2 | 220 | |||
| AF042060.1 | - | 1, 2 | 220 | |||
| AF042061.1 | - | 1, 2 | 220 | |||
| U57439.1 | - | 2, 3 | 636 | |||

Esr2
Genomic Map