siRNA Details
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012758.1 | NP_036890.1 | 4 | 880 | ||
| GenBank mRNA | L20838.1 | - | 2 | 516 | ||
| U21683.1 | - | 4 | 880 | |||
| U21684.1 | - | 4 | 880 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syk| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012758.1 | NP_036890.1 | 4 | 880 | ||
| GenBank mRNA | L20838.1 | - | 2 | 516 | ||
| U21683.1 | - | 4 | 880 | |||
| U21684.1 | - | 4 | 880 | |||