siRNA Details
| Gene Symbol: Vav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012759.2 | NP_036891.2 | 25 | 2452 | ||
| GenBank mRNA | BC091160.1 | - | 25 | 2452 | 7373783 | |
| U39476.1 | - | 25 | 2451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vav1| Gene Symbol: Vav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012759.2 | NP_036891.2 | 25 | 2452 | ||
| GenBank mRNA | BC091160.1 | - | 25 | 2452 | 7373783 | |
| U39476.1 | - | 25 | 2451 | |||