siRNA Details
| Gene Symbol: SLC1A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193493.1 | NP_001180422.1 | 3 | 266 | ||
| NM_003038.4 | NP_003029.2 | 3 | 900 | |||
| XM_006712079.2 | XP_006712142.1 | 3 | 266 | |||
| GenBank mRNA | AB026689.1 | - | 3 | 638 | ||
| AK126636.1 | - | 2 | 2090 | |||
| AK223486.1 | - | 3 | 862 | |||
| AK295687.1 | - | 3 | 221 | |||
| AK298472.1 | - | 4 | 811 | |||
| AK313052.1 | - | 3 | 864 | |||
| AK315986.1 | - | 4 | 822 | |||
| BC026216.1 | - | 3 | 801 | 4420405 | ||
| BC072423.1 | - | 3 | 860 | 6186177 | ||
| DA218921.1 | - | 3 | 268 | |||
| L14595.1 | - | 3 | 800 | |||
| L19444.1 | - | 3 | 779 | |||

SLC1A4
Genomic Map