siRNA Details
| Gene Symbol: Casp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012762.2 | NP_036894.2 | 5 | 730 | ||
| GenBank mRNA | D85899.1 | - | 5 | 730 | ||
| S79676.1 | - | 1 | 73 | |||
| U14647.1 | - | 5 | 730 | |||
| U34621.1 | - | 1 | 49 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casp1| Gene Symbol: Casp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012762.2 | NP_036894.2 | 5 | 730 | ||
| GenBank mRNA | D85899.1 | - | 5 | 730 | ||
| S79676.1 | - | 1 | 73 | |||
| U14647.1 | - | 5 | 730 | |||
| U34621.1 | - | 1 | 49 | |||