siRNA Details
Gene Symbol: Ccnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012766.1 | NP_036898.1 | 2 | 420 | ||
GenBank mRNA | D16309.1 | - | 2 | 420 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012766.1 | NP_036898.1 | 2 | 420 | ||
GenBank mRNA | D16309.1 | - | 2 | 420 |