siRNA Details
| Gene Symbol: Ccnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012766.1 | NP_036898.1 | 2 | 455 | ||
| XM_008766836.1 | XP_008765058.1 | 2 | 232 | |||
| GenBank mRNA | BC085763.1 | - | 2 | 488 | 7189593 | |
| BC089819.1 | - | 645 | 7384331 | |||
| D16309.1 | - | 2 | 455 | |||

Ccnd3
Genomic Map