siRNA Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012767.2 | NP_036899.1 | 2 | 156 | ||
| XM_006252096.2 | XP_006252158.1 | 1 | 157 | |||
| GenBank mRNA | M12579.1 | - | 2 | 156 | ||
| S50870.1 | - | 1 | 130 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnrh1| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012767.2 | NP_036899.1 | 2 | 156 | ||
| XM_006252096.2 | XP_006252158.1 | 1 | 157 | |||
| GenBank mRNA | M12579.1 | - | 2 | 156 | ||
| S50870.1 | - | 1 | 130 | |||