siRNA Details
| Gene Symbol: St6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113344.1 | NP_001106815.1 | 4 | 931 | ||
| NM_147205.2 | NP_671738.2 | 3 | 287 | |||
| XM_006248501.1 | XP_006248563.1 | 5 | 1012 | |||
| XM_008768792.1 | XP_008767014.1 | 2 | 1074 | |||
| GenBank mRNA | FQ210708.1 | - | 3 | 719 | ||
| FQ222565.1 | - | 5 | 1480 | |||
| FQ224146.1 | - | 5 | 1416 | |||
| M18769.1 | - | 4 | 932 | |||
| M83143.1 | - | 3 | 287 | |||

St6gal1
Genomic Map