siRNA Details
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147205.2 | NP_671738.2 | 2, 3 | 237 | ||
GenBank mRNA | M73987.1 | - | 2, 3 | 274 | ||
M83143.1 | - | 2, 3 | 237 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147205.2 | NP_671738.2 | 2, 3 | 237 | ||
GenBank mRNA | M73987.1 | - | 2, 3 | 274 | ||
M83143.1 | - | 2, 3 | 237 |