siRNA Details
| Gene Symbol: Pcsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017091.2 | NP_058787.1 | 11 | 1766 | ||
| GenBank mRNA | M76705.1 | - | 11 | 1768 | ||
| Gene Symbol: LOC100911216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773859.1 | XP_008772081.1 | 10 | 1316 | ||

Pcsk1
Genomic Map